accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 832-10-0

IUPAC Name: cyclotridecanone
Synonyms: cyclotridecanone
Molecular Weight (g/mol): 196.33
Molecular Formula: C13H24O
InChi Key: VHUGWUBIUBBUAF-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCC1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results