accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83654-13-1

IUPAC Name: 2-amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid
Synonyms: AMPA
Molecular Weight (g/mol): 186.17
Molecular Formula: C7H10N2O4
InChi Key: UUDAMDVQRQNNHZ-UHFFFAOYNA-N
SMILES: CC1=C(CC(N)C(O)=O)C(=O)NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results