Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-AMPA, Tocris Bioscience™
SDP

Catalog No. 025310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-531-0 Supplier R&D Systems Supplier No. 0253/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inactive isomer of (RS)-AMPA (Cat. No. 0169)

Inactive enantiomer of AMPA.

Active Enantiomer and Racemate also available.

Chemical Identifiers

CAS 83654-13-1
Molecular Formula C7H10N2O4
Molecular Weight (g/mol) 186.167
InChI Key UUDAMDVQRQNNHZ-RXMQYKEDSA-N
Synonym r-ampa, ampa, d, unii-f45tj02el1, chembl14815, r-2-amino-3-3-hydroxy-5-methyl-isoxazol-4-yl-propionic acid, r-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, d-ampa, --ampa, tocris-0253, biomol-nt_000215
PubChem CID 6604707
IUPAC Name (2R)-2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
SMILES CC1=C(C(=O)NO1)CC(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 186.17
Percent Purity >99%
Chemical Name or Material (R)-AMPA
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.