accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOOHSOOO

CAS RN 83732-66-5

IUPAC Name: sodium 4,6-dihydroxynaphthalene-2-sulfonate
Synonyms: sodium 4,6-dihydroxynaphthalene-2-sulfonate
Molecular Weight (g/mol): 262.21
Molecular Formula: C10H7NaO5S
InChi Key: KJIKTXVDOGQVIJ-UHFFFAOYSA-M
SMILES: [Na+].OC1=CC=C2C=C(C=C(O)C2=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results