accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 838818-26-1

IUPAC Name: 3-(3-{[4-(4-methoxyphenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}propyl)-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 3-(3-{[4-(4-methoxyphenyl)-5-(pyridin-4-yl)-1,2,4-triazol-3-yl]sulfanyl}propyl)-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 521.6
Molecular Formula: C29H23N5O3S
InChi Key: RNUXIZKXJOGYQP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1C(SCCCN2C(=O)C3=CC=CC4=CC=CC(C2=O)=C34)=NN=C1C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results