accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 838823-31-7

IUPAC Name: 4'-[5-({4-[(cyclopropylamino)methyl]phenyl}amino)-2,3-dihydro-1H-pyrazol-3-ylidene]-2-hydroxy-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraen-4-one
Synonyms: 4'-[5-({4-[(cyclopropylamino)methyl]phenyl}amino)-1,2-dihydropyrazol-3-ylidene]-2-hydroxy-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraen-4-one
Molecular Weight (g/mol): 412.49
Molecular Formula: C25H24N4O2
InChi Key: BYFCMDWKTJJQGR-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C=CC1=C1C=CC(C=C1)=C1NNC(NC2=CC=C(CNC3CC3)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results