Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCS 2312, Tocris Bioscience™
SDP

Catalog No. 30381 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-381 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-381 Supplier R&D Systems Supplier No. 3038/1

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent Chk1 inhibitor

Potent and selective checkpoint kinase 1 (chk1) inhibitor (Ki = 0.38 nM, EC50 = 60 nM). Enhances the cell killing activity of Gemcitabine (Cat. No. 3259) in breast and prostate cancer cell lines and displays antiproliferative effects in vitro.

Chemical Identifiers

CAS 838823-31-7
Molecular Formula C25H24N4O2
Molecular Weight (g/mol) 412.493
InChI Key BYFCMDWKTJJQGR-UHFFFAOYSA-N
Synonym 4'-5-3-cyclopropylamino methyl phenyl amino-1h-pyrazol-3-yl-1,1'-biphenyl-2,4-diol, 4-5-4-cyclopropylaminomethyl-phenylamino-2h-pyrazol-3-yl-biphenyl-2,4-diol, tc0
PubChem CID 11258443
IUPAC Name 4-[4-[5-[4-[(cyclopropylamino)methyl]anilino]-1,2-dihydropyrazol-3-ylidene]cyclohexa-2,5-dien-1-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one
SMILES C1CC1NCC2=CC=C(C=C2)NC3=CC(=C4C=CC(=C5C=CC(=O)C=C5O)C=C4)NN3

Specifications

Quantity 1 mg
Formula Weight 412.48
Percent Purity >97%
Chemical Name or Material TCS 2312
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.