accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOCH3

CAS RN 84-72-0

IUPAC Name: ethyl 2-[(2-ethoxy-2-oxoethoxy)carbonyl]benzoate
Synonyms: ethyl carbethoxymethyl phthalate
Molecular Weight (g/mol): 280.28
Molecular Formula: C14H16O6
InChi Key: PZBLUWVMZMXIKZ-UHFFFAOYSA-N
SMILES: CCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results