accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOHOO

CAS RN 84-86-6

IUPAC Name: 4-aminonaphthalene-1-sulfonic acid
Synonyms: naphthionic acid sulfonaphtin 1-naphthylamin-4-sulfonsaeure piria-saeure piria acid usaf M-5 piria's acid kyselina nafthionova 1,4-naphthionic acid naphthionsaeure
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: NRZRRZAVMCAKEP-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)