Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminonaphthalene-1-sulfonic acid, 98%, Thermo Scientific™

Catalog No. AAB2182922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2182922 100 g
AAB2182914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2182922 Supplier Thermo Scientific Chemicals Supplier No. B2182922

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Amino-1-naphthalenesulfonic acid has potent therapeutic applications. Its sodium salt has been used as non-toxic hemostatic. It is also a metabolite of azo dyes.

Solubility
Sparingly soluble in dichloromethane. Soluble in aqueous base.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-86-6
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00004027
InChI Key NRZRRZAVMCAKEP-UHFFFAOYSA-N
Synonym naphthionic acid, 4-amino-1-naphthalenesulfonic acid, piria's acid, 1-naphthylamine-4-sulfonic acid, naphthionsaeure, 1,4-naphthionic acid, sulfonaphtin, 1-amino-4-sulfonaphthalene, usaf m-5, 1-aminonaphthalene-4-sulfonic acid
PubChem CID 6790
ChEBI CHEBI:38219
IUPAC Name 4-aminonaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Melting Point >300°C
Quantity 100 g
UN Number UN2585
Beilstein 1971299
Merck Index 14,6403
Solubility Information Sparingly soluble in dichloromethane. Soluble in aqueous base.
Formula Weight 223.25
Percent Purity 98%
Chemical Name or Material 4-Aminonaphthalene-1-sulfonic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 201-567-9
RTECSNumber QK1270000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.