Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-naphthalenesulfonic Acid 98%, Thermo Scientific™

Catalog No. AC400820250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400820250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC400820250 Supplier Thermo Scientific Chemicals Supplier No. 400820250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-86-6
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00004027
InChI Key NRZRRZAVMCAKEP-UHFFFAOYSA-N
Synonym naphthionic acid, 4-amino-1-naphthalenesulfonic acid, piria's acid, 1-naphthylamine-4-sulfonic acid, naphthionsaeure, 1,4-naphthionic acid, sulfonaphtin, 1-amino-4-sulfonaphthalene, usaf m-5, 1-aminonaphthalene-4-sulfonic acid
PubChem CID 6790
ChEBI CHEBI:38219
IUPAC Name 4-aminonaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
Merck Index 15, 6485
Formula Weight 223.25
Percent Purity ≥97.5%
Physical Form Powder or Granules
Chemical Name or Material 4-Amino-1-naphthalenesulfonic acid, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement AIR SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.