accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHONCH3O

CAS RN 84088-42-6

IUPAC Name: 2-hydroxy-N,1-dimethyl-4-oxo-N-phenyl-1,4-dihydroquinoline-3-carboxamide
Synonyms: roquinimex
Molecular Weight (g/mol): 308.34
Molecular Formula: C18H16N2O3
InChi Key: ONCYIVQBAJEKOT-UHFFFAOYSA-N
SMILES: CN(C(=O)C1=C(O)N(C)C2=CC=CC=C2C1=O)C1=CC=CC=C1
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)