Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linomide 98.0+%, TCI America™
SDP

Catalog No. L027010MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L027010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L027010MG Supplier TCI America Supplier No. L027010MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84088-42-6
Molecular Formula C18H16N2O3
Molecular Weight (g/mol) 308.337
MDL Number MFCD00866331
InChI Key SGOOQMRIPALTEL-UHFFFAOYSA-N
Synonym linomide, roquinimex, roquinimexum latin, roquinimex usan:inn, ccris 7673, fcf 89, fcf-89, n-phenyl-n-methyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxo-quinoline-3-carboxamide, 1,2-dihydro-4-hydroxy-n,1-dimethyl-2-oxo-3-quinolinecarboxanilide, 4-hydroxy-n,1-dimethyl-2-oxo-n-phenyl-1,2-dihydroquinoline-3-carboxamide
PubChem CID 54676478
IUPAC Name 4-hydroxy-N,1-dimethyl-2-oxo-N-phenylquinoline-3-carboxamide
SMILES CN1C2=CC=CC=C2C(=C(C1=O)C(=O)N(C)C3=CC=CC=C3)O

Specifications

Melting Point 204°C
Color White
Quantity 10 mg
Formula Weight 308.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Linomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.