Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linomide

Catalog No. AAJ65453MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65453MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65453MA Supplier Thermo Scientific Chemicals Supplier No. J65453MA
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 84088-42-6 | C18H16N2O3 | 308.337 g/mol

An immunomodulator with antitumour properties. Inhibits T-cell apoptosis; also inhibits angiogenesis in rats

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunomodulator with antitumour properties
  • Inhibits T-cell apoptosis; also inhibits angiogenesis in rats
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84088-42-6
Molecular Formula C18H16N2O3
Molecular Weight (g/mol) 308.337
MDL Number MFCD00866331
InChI Key SGOOQMRIPALTEL-UHFFFAOYSA-N
Synonym linomide, roquinimex, roquinimexum latin, roquinimex usan:inn, ccris 7673, fcf 89, fcf-89, n-phenyl-n-methyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxo-quinoline-3-carboxamide, 1,2-dihydro-4-hydroxy-n,1-dimethyl-2-oxo-3-quinolinecarboxanilide, 4-hydroxy-n,1-dimethyl-2-oxo-n-phenyl-1,2-dihydroquinoline-3-carboxamide
PubChem CID 54676478
IUPAC Name 4-hydroxy-N,1-dimethyl-2-oxo-N-phenylquinoline-3-carboxamide
SMILES CN1C2=CC=CC=C2C(=C(C1=O)C(=O)N(C)C3=CC=CC=C3)O

Specifications

Color White
Quantity 10 mg
Formula Weight 308.33
Physical Form Solid
Chemical Name or Material Linomide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.