accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOOOCH3OOH3COOCH3OOCH3OHOOCH3

CAS RN 84380-10-9

IUPAC Name: methyl 4-(acetyloxy)-5-acetamido-2-hydroxy-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Synonyms: methyl 4-(acetyloxy)-5-acetamido-2-hydroxy-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Molecular Weight (g/mol): 491.45
Molecular Formula: C20H29NO13
InChi Key: CAKDMYCBPKFKCV-UHFFFAOYNA-N
SMILES: COC(=O)C1(O)CC(OC(C)=O)C(NC(C)=O)C(O1)C(OC(C)=O)C(COC(C)=O)OC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results