accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNH2S

CAS RN 84544-86-5

IUPAC Name: [1-amino-2-(phenylsulfanyl)ethylidene]azanium
Synonyms: [1-amino-2-(phenylsulfanyl)ethylidene]azanium
Molecular Weight (g/mol): 167.25
Molecular Formula: C8H11N2S
InChi Key: QTFCJAKFFGFURL-UHFFFAOYSA-O
SMILES: NC(=[NH2+])CSC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results