accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84672-48-0

IUPAC Name: 2-bromophenyl 4-methylbenzene-1-sulfonate
Synonyms: 2-bromophenyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 327.19
Molecular Formula: C13H11BrO3S
InChi Key: ZXJVRRGBOXUFSQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results