accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 847264-30-6

IUPAC Name: 3-(2-bromophenyl)pyridine
Synonyms: 3-(2-bromophenyl)pyridine
Molecular Weight (g/mol): 234.1
Molecular Formula: C11H8BrN
InChi Key: XEQMSSLGEYCVDC-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CN=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results