accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNONHFOO

CAS RN 847553-89-3

IUPAC Name: N-[(4-fluorophenyl)methyl]-3-oxo-1,1-diphenyl-hexahydro-1H-[1,3]oxazolo[3,4-a]pyrazine-7-carboxamide
Synonyms: N-[(4-fluorophenyl)methyl]-3-oxo-1,1-diphenyl-tetrahydro-[1,3]oxazolo[3,4-a]pyrazine-7-carboxamide
Molecular Weight (g/mol): 445.49
Molecular Formula: C26H24FN3O3
InChi Key: SFRQIPRTNYHJHP-UHFFFAOYNA-N
SMILES: FC1=CC=C(CNC(=O)N2CCN3C(C2)C(OC3=O)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)