accessibility menu, dialog, popup

UserName

Viewing profile as user:
CNH2CNH2HClPdClHPd

CAS RN 847616-85-7

IUPAC Name: (chloraniumylpalladio)chloranium bis(2'-amino-[1,1'-biphenyl]-2-ide) palladium
Synonyms: (chloraniumylpalladio)chloranium bis(2'-amino-[1,1'-biphenyl]-2-ide) palladium
Molecular Weight (g/mol): 622.19
Molecular Formula: C24H22Cl2N2Pd2
InChi Key: GYIYBDXOIBSKGQ-UHFFFAOYSA-N
SMILES: [Pd].[ClH+][Pd][ClH+].NC1=CC=CC=C1C1=CC=CC=[C-]1.NC1=CC=CC=C1C1=CC=CC=[C-]1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)