accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 84976-67-0

IUPAC Name: 4'-ethyl-[1,1'-bi(cyclohexane)]-4-carboxylic acid
Synonyms: 4'-ethyl-[1,1'-bi(cyclohexane)]-4-carboxylic acid
Molecular Weight (g/mol): 238.37
Molecular Formula: C15H26O2
InChi Key: OQIHEFMTIUJJET-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1CCC(CC1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results