accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85030-56-4

IUPAC Name: 2,5,8,11-tetraoxatridecan-13-amine
Synonyms: 2,5,8,11-tetraoxatridecan-13-amine
Molecular Weight (g/mol): 207.27
Molecular Formula: C9H21NO4
InChi Key: DQTQYVYXIOQYGN-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCN
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)