Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6,9,12-Tetraoxatridecanamine 98.0+%, TCI America™
SDP

Catalog No. M2501100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2501100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2501100MG Supplier TCI America Supplier No. M2501100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85030-56-4
Molecular Formula C9H21NO4
Molecular Weight (g/mol) 207.27
MDL Number MFCD11041099
InChI Key DQTQYVYXIOQYGN-UHFFFAOYSA-N
Synonym Methyl-PEG4-Amine, mPEG4-NH2, mPEG4-Amine
PubChem CID 22639371
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanamine
SMILES COCCOCCOCCOCCN

Specifications

Color Yellow
Boiling Point 85°C
Quantity 100 mg
UN Number 2735
Formula Weight 207.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,6,9,12-Tetraoxatridecanamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.