accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850568-25-1

IUPAC Name: {4-[(pyridin-2-yl)carbamoyl]phenyl}boronic acid
Synonyms: 4-[(pyridin-2-yl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 242.04
Molecular Formula: C12H11BN2O3
InChi Key: LEAZDTWQWNXKJM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)NC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results