accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850568-72-8

BOOH3CCH3H3CH3COOCH3H3CCH3

CAS RN 850568-72-8

IUPAC Name: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 304.19
Molecular Formula: C17H25BO4
InChi Key: GNXLDEFJAZGNCJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)