Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97.0+%, TCI America™
SDP

Catalog No. B51325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51325G 5 g
B51321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51325G Supplier TCI America Supplier No. B51325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 850568-72-8
Molecular Formula C17H25BO4
Molecular Weight (g/mol) 304.19
MDL Number MFCD03453055
InChI Key GNXLDEFJAZGNCJ-UHFFFAOYSA-N
Synonym tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-tert-butoxycarbonyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonyl benzeneboronic acid pinacol ester, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-tert-butoxycarbonyl phenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid tert-butyl ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid tert-butyl ester, 4-boc-phenylboronic acid pinacol ester, 4-tert-butoxycarbonyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonyl phenyl boronic acid pinacol ester
PubChem CID 4376766
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES CC(C)(C)OC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 142°C
Color White
Quantity 5 g
Formula Weight 304.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.