accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 850664-21-0

IUPAC Name: N-(3-{[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-1H-imidazo[4,5-c]pyridin-6-yl]oxy}phenyl)-4-[2-(morpholin-4-yl)ethoxy]benzamide
Synonyms: N-(3-{[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-6-yl]oxy}phenyl)-4-[2-(morpholin-4-yl)ethoxy]benzamide
Molecular Weight (g/mol): 570.61
Molecular Formula: C29H30N8O5
InChi Key: YOVNFNXUCOWYSG-UHFFFAOYSA-N
SMILES: CCN1C2=CC(OC3=CC=CC(NC(=O)C4=CC=C(OCCN5CCOCC5)C=C4)=C3)=NC=C2N=C1C1=NON=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)