accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85167-10-8

IUPAC Name: 4-butyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0²,⁶]decane
Synonyms: 4-butyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0²,⁶]decane
Molecular Weight (g/mol): 236.16
Molecular Formula: C14H25BO2
InChi Key: IMUJPBOUZJFWJB-UHFFFAOYNA-N
SMILES: CCCCB1OC2CC3CC(C3(C)C)C2(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results