accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 852054-42-3

IUPAC Name: 7-bromo-2H,3H-thieno[3,4-b][1,4]dioxine-5-carbaldehyde
Synonyms: 7-bromo-2H,3H-thieno[3,4-b][1,4]dioxine-5-carbaldehyde
Molecular Weight (g/mol): 249.08
Molecular Formula: C7H5BrO3S
InChi Key: YEKBJVHBVICUOZ-UHFFFAOYSA-N
SMILES: BrC1=C2OCCOC2=C(S1)C=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)