Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde 96.0+%, TCI America™
SDP

Catalog No. B5396200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5396200MG 200 mg
B53961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5396200MG Supplier TCI America Supplier No. B5396200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 852054-42-3
Molecular Formula C7H5BrO3S
Molecular Weight (g/mol) 249.078
MDL Number MFCD08690306
InChI Key YEKBJVHBVICUOZ-UHFFFAOYSA-N
Synonym 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, 2,3-dihydro-7-bromothieno 3,4-b 1,4 dioxin-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 7-bromo-2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde,7-bromo-2,3-dihydro
PubChem CID 11391008
IUPAC Name 5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-7-carbaldehyde
SMILES C1COC2=C(SC(=C2O1)C=O)Br

Specifications

Melting Point 149°C
Color Gray-White
Quantity 200 mg
Formula Weight 249.08
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.