accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNS

CAS RN 852180-66-6

IUPAC Name: methyl({[2-(thiophen-2-yl)phenyl]methyl})amine
Synonyms: methyl({[2-(thiophen-2-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 203.3
Molecular Formula: C12H13NS
InChi Key: MRKJJEJYTBOUTH-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results