Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(2-thien-2-ylbenzyl)amine, 97%, Thermo Scientific™

Catalog No. CC35546DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC35546DE 5 g
CC35546CB 250 mg
CC35546DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC35546DE Supplier Thermo Scientific Chemicals Supplier No. CC35546DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 852180-66-6
Molecular Formula C12H13NS
Molecular Weight (g/mol) 203.303
MDL Number MFCD07368538
InChI Key MRKJJEJYTBOUTH-UHFFFAOYSA-N
Synonym n-methyl-n-2-thien-2-ylbenzyl amine, n-methyl-1-2-thiophen-2-ylphenyl methanamine, n-methyl-2-thien-2-ylbenzylamine, methyl 2-thiophen-2-yl phenyl methyl amine, n-methyl-1-2-thiophen-2-yl phenyl methanamine, d0a2yg, n-methyl-n-2-thiophen-2-ylbenzyl amine, methyl 2-2-thienyl phenyl methyl amine, benzenemethanamine,n-methyl-2-2-thienyl, benzenemethanamine, n-methyl-2-2-thienyl
PubChem CID 7060565
IUPAC Name N-methyl-1-(2-thiophen-2-ylphenyl)methanamine
SMILES CNCC1=CC=CC=C1C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 203.31
Percent Purity 95%
Chemical Name or Material N-methyl-N-(2-thien-2-ylbenzyl)amine
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.