accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOOHOH2N

CAS RN 852679-66-4

IUPAC Name: 2-amino-4-methylbicyclo[3.1.0]hexane-2,6-dicarboxylic acid
Synonyms: 2-amino-4-methylbicyclo[3.1.0]hexane-2,6-dicarboxylic acid
Molecular Weight (g/mol): 199.21
Molecular Formula: C9H13NO4
InChi Key: BQVZICWQBFTOJX-UHFFFAOYNA-N
SMILES: CC1CC(N)(C2C1C2C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results