Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-LY 395756, Tocris Bioscience™

Catalog No. 327210
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-7210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-7210 Supplier R&D Systems Supplier No. 3272/10
This item is not returnable. View return policy

Mixed mGlu2 agonist/ mGlu3 antagonist

Selective metabotropic glutamate ligand for mGlu2 and mGlu3 receptors (Ki values are 0.165 μM and 0.302 μM respectively). Acts as an agonist at mGlu2 and antagonist at mGlu3. Analog of LY 354740 (Cat. No. 3246).

Chemical Identifiers

CAS 852679-66-4
Molecular Formula C9H13NO4
Molecular Weight (g/mol) 199.206
InChI Key BQVZICWQBFTOJX-DKNAPFGQSA-N
Synonym 1s,2s,4r,5r,6s-rel-2-amino-4-methylbicyclo 3.1.0 hexane-2,6-dicarboxylic acid
PubChem CID 90488822
IUPAC Name (1S,2S,4R,6S)-2-amino-4-methylbicyclo[3.1.0]hexane-2,6-dicarboxylic acid
SMILES CC1CC(C2C1C2C(=O)O)(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 199.21
Percent Purity >98%
Chemical Name or Material (±)-LY 395756
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.