accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85272-31-7

IUPAC Name: di-tert-butyl(trifluoromethanesulfonyloxy)silyl trifluoromethanesulfonate
Synonyms: di-tert-butyl(trifluoromethanesulfonyloxy)silyl trifluoromethanesulfonate
Molecular Weight (g/mol): 440.44
Molecular Formula: C10H18F6O6S2Si
InChi Key: HUHKPYLEVGCJTG-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](OS(=O)(=O)C(F)(F)F)(OS(=O)(=O)C(F)(F)F)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)