CAS RN 85375-15-1
IUPAC Name:
1-(4,4-diphenylbut-3-en-1-yl)piperidine-3-carboxylic acid hydrochloride
Synonyms:
1-(4,4-diphenylbut-3-en-1-yl)piperidine-3-carboxylic acid hydrochloride
Molecular Weight (g/mol):
371.91
Molecular Formula:
C22H26ClNO2
InChi Key:
SNGGBKYQZVAQKA-UHFFFAOYNA-N
SMILES:
Cl.OC(=O)C1CCCN(CCC=C(C2=CC=CC=C2)C2=CC=CC=C2)C1
1
–
1
of
1
results
1
R&D Systems™ SKF 89976A hydrochloride
CAS: 85375-15-1 Molecular Formula: C22H26ClNO2