accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85614-43-3

IUPAC Name: methyl 2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzoate
Synonyms: methyl 2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzoate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: LYSUGZLJKRSLHM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(CC=C)=CC(OC)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results