Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Allyl-3-methoxysalicylate 98.0+%, TCI America™
SDP

Catalog No. M220425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M220425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M220425G Supplier TCI America Supplier No. M220425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85614-43-3
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD04038819
InChI Key LYSUGZLJKRSLHM-UHFFFAOYSA-N
Synonym 5-Allyl-3-methoxysalicylic Acid Methyl Ester, Methyl 5-Allyl-2-hydroxy-3-methoxybenzoate, 5-Allyl-2-hydroxy-3-methoxybenzoic Acid Methyl Ester
PubChem CID 3020842
IUPAC Name methyl 2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzoate
SMILES COC(=O)C1=CC(CC=C)=CC(OC)=C1O

Specifications

Melting Point 56°C
Color White
Boiling Point 174°C
Quantity 25 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Allyl-3-methoxysalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.