accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857290-04-1

IUPAC Name: N-(6-aminopyridin-2-yl)-4'-cyano-[1,1'-biphenyl]-4-sulfonamide
Synonyms: N-(6-aminopyridin-2-yl)-4'-cyano-[1,1'-biphenyl]-4-sulfonamide
Molecular Weight (g/mol): 350.4
Molecular Formula: C18H14N4O2S
InChi Key: ZESFDAKNYJQYKO-UHFFFAOYSA-N
SMILES: NC1=NC(NS(=O)(=O)C2=CC=C(C=C2)C2=CC=C(C=C2)C#N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results