accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85737-06-0

IUPAC Name: 1-(2-bromo-1,1,2,2-tetrafluoroethoxy)-1,2,2-trifluoroethene
Synonyms: 1-(2-bromo-1,1,2,2-tetrafluoroethoxy)-1,2,2-trifluoroethene
Molecular Weight (g/mol): 276.94
Molecular Formula: C4BrF7O
InChi Key: ZPYGRBTUNITHKJ-UHFFFAOYSA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(F)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results