Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromotetrafluoroethyl Trifluorovinyl Ether (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. B41695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41695G Supplier TCI America Supplier No. B41695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85737-06-0
Molecular Formula C4BrF7O
Molecular Weight (g/mol) 276.936
MDL Number MFCD02183517
InChI Key ZPYGRBTUNITHKJ-UHFFFAOYSA-N
PubChem CID 2782267
IUPAC Name 1-bromo-1,1,2,2-tetrafluoro-2-(1,2,2-trifluoroethenoxy)ethane
SMILES C(=C(F)F)(OC(C(F)(F)Br)(F)F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 276.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromotetrafluoroethyl Trifluorovinyl Ether (stabilized with MEHQ)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.