accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHONHNH

CAS RN 85753-43-1

IUPAC Name: 3,13,23-triazahexacyclo[14.7.0.0²,¹⁰.0⁴,⁹.0¹¹,¹⁵.0¹⁷,²²]tricosa-1(16),2(10),4,6,8,11(15),17(22),18,20-nonaen-12-one
Synonyms: 3,13,23-triazahexacyclo[14.7.0.0²,¹⁰.0⁴,⁹.0¹¹,¹⁵.0¹⁷,²²]tricosa-1(16),2(10),4,6,8,11(15),17(22),18,20-nonaen-12-one
Molecular Weight (g/mol): 311.34
Molecular Formula: C20H13N3O
InChi Key: MEXUTNIFSHFQRG-UHFFFAOYSA-N
SMILES: O=C1NCC2=C1C1=C(NC3=CC=CC=C13)C1=C2C2=C(N1)C=CC=C2
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results