accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNH2

CAS RN 857549-15-6

IUPAC Name: 1-(4-methyl-1,3-thiazol-5-yl)methanamine
Synonyms: 1-(4-methyl-1,3-thiazol-5-yl)methanamine
Molecular Weight (g/mol): 128.19
Molecular Formula: C5H8N2S
InChi Key: UGNFDYLWMWQMPR-UHFFFAOYSA-N
SMILES: CC1=C(CN)SC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results