CAS RN 85798-08-9
IUPAC Name:
hydrogen (4aR,8aR)-5-propyl-1H,4H,4aH,5H,6H,7H,8H,8aH,9H-pyrido[2,3-f]indazole chloride
Synonyms:
hydrogen quinpirole chloride
Molecular Weight (g/mol):
255.79
Molecular Formula:
C13H22ClN3
InChi Key:
HJHVRVJTYPKTHX-HTMVYDOJSA-N
SMILES:
[H+].[Cl-].CCCN1CCC[C@@H]2CC3=C(C[C@@H]12)C=NN3
1
–
1
of
1
results
1
R&D Systems™ (-)-Quinpirole hydrochloride
CAS: 85798-08-9 Molecular Formula: C13H22ClN3