accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrI

CAS RN 858841-53-9

IUPAC Name: 1-bromo-2-iodo-4-methylbenzene
Synonyms: 1-bromo-2-iodo-4-methylbenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: FLOUOTRVXNQGKP-UHFFFAOYSA-N
SMILES: CC1=CC(I)=C(Br)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results