CAS RN 859502-43-5
IUPAC Name:
[1-(2-methanesulfonamidoethyl)piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate; sulfamic acid
Synonyms:
[1-(2-methanesulfonamidoethyl)piperidin-4-yl]methyl 5-fluoro-2-methoxy-1H-indole-3-carboxylate; sulfamic acid
Molecular Weight (g/mol):
524.58
Molecular Formula:
C19H29FN4O8S2
InChi Key:
VQJFDBXITIOGJM-UHFFFAOYSA-N
SMILES:
NS(O)(=O)=O.COC1=C(C(=O)OCC2CCN(CCNS(C)(=O)=O)CC2)C2=CC(F)=CC=C2N1
1
–
1
of
1
results
1
R&D Systems™ GR 125487 sulfamate
CAS: 859502-43-5 Molecular Formula: C19H29FN4O8S2