CAS RN 859850-67-2
IUPAC Name:
{[4-(furan-2-yl)phenyl]methyl}(methyl)amine
Synonyms:
{[4-(furan-2-yl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol):
187.24
Molecular Formula:
C12H13NO
InChi Key:
QJHXIMBULPYCDO-UHFFFAOYSA-N
SMILES:
CNCC1=CC=C(C=C1)C1=CC=CO1
1
–
1
of
1
results
1
N-[4-(2-Furyl)benzyl]-N-methylamine, ≥95%, Thermo Scientific™
CAS: 859850-67-2 Molecular Formula: C12H13NO Molecular Weight (g/mol): 187.24 MDL Number: MFCD08060507 InChI Key: QJHXIMBULPYCDO-UHFFFAOYSA-N Synonym: n-4-2-furyl benzyl-n-methylamine,4-furan-2-yl phenyl methyl methyl amine,benzenemethanamine,4-2-furanyl-n-methyl,1-4-furan-2-yl phenyl-n-methylmethanamine,4-2-furyl phenyl methyl methylamine PubChem CID: 7537512 IUPAC Name: {[4-(furan-2-yl)phenyl]methyl}(methyl)amine SMILES: CNCC1=CC=C(C=C1)C1=CC=CO1