CAS RN 86-92-0
IUPAC Name:
3-methyl-1-(4-methylphenyl)-4,5-dihydro-1H-pyrazol-5-one
Synonyms:
5-methyl-2-(4-methylphenyl)-4H-pyrazol-3-one
Molecular Weight (g/mol):
188.23
Molecular Formula:
C11H12N2O
InChi Key:
IOQOLGUXWSBWHR-UHFFFAOYSA-N
SMILES:
CC1=NN(C(=O)C1)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
3-Methyl-1-p-tolyl-5-pyrazolone 98.0+%, TCI America™
CAS: 86-92-0 Molecular Formula: C11H12N2O Molecular Weight (g/mol): 188.23 MDL Number: MFCD00035708 InChI Key: IOQOLGUXWSBWHR-UHFFFAOYSA-N PubChem CID: 66591 IUPAC Name: 3-methyl-1-(4-methylphenyl)-4,5-dihydro-1H-pyrazol-5-one SMILES: CC1=NN(C(=O)C1)C1=CC=C(C)C=C1