accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONH

CAS RN 86-99-7

IUPAC Name: 7-chloro-1,4-dihydroquinolin-4-one
Synonyms: 7-chloro-1H-quinolin-4-one
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: XMFXTXKSWIDMER-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=O)C=CNC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results