accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86303-22-2

IUPAC Name: 2,3,4,5,6-pentahydroxy-N-(3-{N-[3-(2,3,4,5,6-pentahydroxyhexanamido)propyl]-4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido}propyl)hexanamide
Synonyms: 2,3,4,5,6-pentahydroxy-N-(3-{N-[3-(2,3,4,5,6-pentahydroxyhexanamido)propyl]-4-{4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido}propyl)hexanamide
Molecular Weight (g/mol): 878.07
Molecular Formula: C42H75N3O16
InChi Key: ZWEVPYNPHSPIFU-UHFFFAOYNA-N
SMILES: CC(CCC(=O)N(CCCNC(=O)C(O)C(O)C(O)C(O)CO)CCCNC(=O)C(O)C(O)C(O)C(O)CO)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results